Starch, soluble (from potato)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9005-84-9
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026
:162.14 g/mol
Merck Index:14,8799
Melting Pt:256…258 °C (dec.)
Storage Temperature:Ambient
EINECS:232-686-4