Anisole 99%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-66-3
:
Formula:C₇H₈O
MDL Number:MFCD00008354
:108.14 g/mol
Merck Index:13,00672
Boiling Pt:154 °C (760 mmHg)
Melting Pt:–37 °C
Density:0.99 g/cm³
Flash Pt:45 °C
Storage Temperature:Ambient
EINECS:202-876-1
UN:2222
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5160 to 1.5190 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥98.5 %