


1-Chloro-2-nitrobenzene 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:88-73-3
:
Formula:C₆H₄ClNO₂
MDL Number:MFCD00007061
:157.56 g/mol
Merck Index:13,02170
Boiling Pt:246 °C (1013 hPa)
Melting Pt:32…34 °C
Density:1.348 g/cm³
Flash Pt:124 °C
Storage Temperature:Ambient
EINECS:201-854-9
UN:1578
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Yellow Fused crystalline mass
- Infrared spectrum:Conforms
- Melting point:32 to 34 °C
- GC:≥99.0 %