Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hjem
2-Chloro-1,4-difluorobenzene
2-Chloro-1,4-difluorobenzene
2-Chloro-1,4-difluorobenzene
2-Chloro-1,4-difluorobenzene
Leverandør:
Thermo Scientific
Sertifikat
Expand image
2-Chloro-1,4-difluorobenzene
Leverandør:
Thermo Scientific
Om dette produktet
CAS-nummer:
2367-91-1
:
Formel:
C₆H₃ClF₂
MDL-nummer:
MFCD00042571
:
148.54 g/mol
Kokende Pt:
128 °C (1013 hPa)
Molten Pt:
–24,6 °C
Tetthet:
1,3561 g/cm³ (20 °C)