
2,5-Dichlorobenzoic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:50-79-3
:
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD00002416
:191.01 g/mol
Boiling Pt:301 °C (760 mmHg)
Melting Pt:152…157 °C
Storage Temperature:Ambient
EINECS:200-065-7
:GHS07
Specification Test Results
- Appearance:White to beige to light brown Powder
- Infrared spectrum:Conforms
- Melting point:152 to 157 °C
- HPLC:≥96.0 %
- Titration with NaOH:≥96.0 %