Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-6-nitrobenzonitrile
2-Chloro-6-nitrobenzonitrile ≥98%
2-Chloro-6-nitrobenzonitrile ≥98%
:
Thermo Fisher Scientific
2-Chloro-6-nitrobenzonitrile ≥98%
:
Thermo Fisher Scientific
:
6575-07-1
:
:
C₇H₃ClN₂O₂
:
MFCD00007203
:
182.57 g/mol
:
118…124 °C
:
229-496-9
:
GHS07
:
Ambient
msds_ALFAA12703
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years