

4-Chlorobenzaldehyde for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:104-88-1
:
Formula:C₇H₅ClO
MDL Number:MFCD00003379
:140.57 g/mol
Boiling Pt:214 °C (1013 hPa)
Melting Pt:46 °C
Density:1.196 g/cm³ (20 °C)
Flash Pt:88 °C
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
EINECS:203-247-4
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:935 mg/l (20 °C)
- Melting Point:46 °C
- Molar Mass:140.57 g/mol
- Boiling Point:214 °C (1013 hPa)
- Vapor Pressure:0.36 hPa (25 °C)
- Flash Point:88 °C
- Density:1.19 g/cm3 (60 °C)
- Ignition Point:302 °C