Order Entry
Norway
ContactUsLinkComponent
1,4-Dichlorobenzene for synthesis, Sigma-Aldrich®
Health
Irritant
Environment
1,4-Dichlorobenzene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Health
Irritant
Environment
1,4-Dichlorobenzene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.08 g/l (25 °C)
  • Melting Point:
    52 - 54 °C
  • Molar Mass:
    147 g/mol
  • Bulk Density:
    800 kg/m3
  • Boiling Point:
    173 - 175 °C (1013 hPa)
  • Vapor Pressure:
    0.8 hPa (20 °C)
  • Flash Point:
    66 °C
  • Refractive Index:
    1.5267 (70 °C, 589 nm)
  • Explosion Limit:
    1.7 - 5.9 %(V)
  • Density:
    1.23 g/cm3 (70 °C)
  • pH:
    7 (0.06 g/l, H2O, 20 °C)
  • Ignition Point:
    500 °C
  • Saturation concentration:
    5 g/m3 (20 °C) Air
  • Hazard Class:
    9
  • Hazard Class Description:
    Miscellaneous Hazardous Material

Specification Test Results

Related Information

Frequently Bought Together