VWR®, Mats, Cleanroom Anti-Fatigue, Bubble and Smooth
Supplier: VWR Collection
Certificates
About this item
These mats offer the comfort of a polyurethane mat which is suitable for ISO Class 5 cleanroom (Class 100) environments. It is either available with a bubble surface, that features a unique ergonomic and anti-fatigue design, or with a smooth surface that is ideal for areas that combine sitting and standing.
- Extremely good chemical and UV resistance (except for organic solvents)
- Free from silicone and latex
- Closed cell/non porous surface allows for easy wash-down and cleaning
- Smooth mat: Ideal if carts or trolleys must be used
- Bubble mat: Ergonomic benefit of stimulating the body to increase blood circulation
- Warranty up to 12 years
DIN 53505, EN ISO 14664-1
Certifications: DIN 53505, EN ISO 14664-1