Bottles, wide neck, with tamper evident cap
Supplier: KARTELL
Certificates
About this item
HDPE, translucent, with black screw caps.
- Good chemical resistance
- Good temperature resistance
The tamper-evident cap incorporates a safety ring which is secured when the cap is screwed on to the bottle. Opening the bottle breaks the safety ring, showing signs of interference.