Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tri-Sodium phosphate dodecahydrate
Trisodium phosphate dodecahydrate
Trisodium phosphate dodecahydrate
Trisodium phosphate dodecahydrate
SWEDTR932403
:
SWED HANDLING
:
Trisodium phosphate dodecahydrate
SWEDTR932403
:
SWED HANDLING
:
Pk:
25 kg
Seq:
4283
:
:
380.12 g/mol
:
13,08736
:
∼75 °C
:
1.62 g/cm³ (20 °C)
:
Ambient
:
231-509-8
:
8,III
:
01-2119489800-32-xxxx
:
GHS07
:
10101-89-0
:
Na₃PO₄·12H₂O
:
MFCD00149198