Order Entry
Norway
ContactUsLinkComponent
Sodium sulphide hydrate
Sodium sulphide hydrate
Catalog # SWEDNS804459
Supplier:  SWED HANDLING
CAS Number:  
Sodium sulphide hydrate
Catalog # SWEDNS804459
Supplier:  SWED HANDLING
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :96.06 g/mol (anhy)
Merck Index:12,08829
Boiling Pt:∼920 °C (1013 hPa)
Melting Pt:50 °C
Density:∼1.43 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:215-211-5
UN:1849
ADR:8,II
REACH:01-2119513694-38
 :GHS06
 :GHS05
 :GHS09
CAS Number:27610-45-3
Formula:Na₂S·xH₂O
MDL Number:MFCD00149183