Order Entry
Norway
ContactUsLinkComponent
Aluminium sulphate
Aluminium sulphate
Catalog # SWEDAL902403
Supplier:  SWED HANDLING
CAS Number:  
Aluminium sulphate
Catalog # SWEDAL902403
Supplier:  SWED HANDLING
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :342.15 g/mol
Merck Index:12,00381
Melting Pt:770 °C
Density:2.672 g/cm³ (22.5 °C)
EINECS:233-135-0
 :GHS05
CAS Number:10043-01-3
Formula:Al₂O₁₂S₃
MDL Number:MFCD00003423