Order Entry
Norway
ContactUsLinkComponent
 
Organic reference standard, Methyl methacrylate (monomer) 1,000 µg/ml in methanol, Supelco®
  40439.
 :  Merck
 :  
Organic reference standard, Methyl methacrylate (monomer) 1,000 µg/ml in methanol, Supelco®
  40439.
 :  Merck
 :  

 

  • Pk:
    1
  • CAS No.:
    80-62-6
  • Description:
    Methyl methacrylate (monomer) 1,000 µg/ml in methanol
  • Formula:
    C₅H₈O₂
  • Description:
    Methyl methacrylate (monomer)