Sulpholane, Supelco®
Catalog # 88965-5ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:120.17 g/mol
:M-Clarity™ quality level = MQ100
EINECS:204-783-1
:GHS07
CAS Number:126-33-0
Formula:C₄H₈O₂S
MDL Number:MFCD00005484