Order Entry
Norway
ContactUsLinkComponent
 
2,4-Diamino-6-phenyl-1,3,5-triazine, Sigma-Aldrich®
 :  Merck
 
2,4-Diamino-6-phenyl-1,3,5-triazine, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    0.3 g/l (25 °C)
  • Melting Point:
    224 - 227 °C
  • Molar Mass:
    187.21 g/mol
  • Bulk Density:
    680 kg/m3
  • Vapor Pressure:
    (20 °C)
  • Flash Point:
    270 °C
  • Density:
    1.425 g/cm3 (15 °C)
  • pH:
    6.5 (0.3 g/l, H2O, 20 °C)