1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate, Millipore®
Katalog # 1.01922.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:1350 g/l (20 °C)
- Melting Point:300 °C
- Molar Mass:332.22 g/mol
- Bulk Density:465 kg/m3
- pH:5.0 (10 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:332.22 g/mol
:M-Clarity™ quality level = MQ200
EINECS:205-741-5
:GHS07
Storage Temperature:Ambient
CAS Number:149-45-1
:MFCD00007473
Formula:C₆H₄Na₂O₈S₂·1H₂O
MDL Number:MFCD00149531