Order Entry
Norway
ContactUsLinkComponent
Organic reference standard, 3-Ethylphenol 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, 3-Ethylphenol 100 µg/ml in methanol, Dr Ehrenstorfer
Katalog # EHERXA13351000ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 3-Ethylphenol 100 µg/ml in methanol, Dr Ehrenstorfer
Katalog # EHERXA13351000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    620-17-7
  • Description:
    3-Ethylphenol 100 µg/ml in methanol
  • Formula:
    C₈H₁₀O
  • Description:
    3-Ethylphenol

Specifications

About this item