Organic reference standard, 4-Chloro-o-toluidine 100 µg/ml in methanol, Dr Ehrenstorfer
EHERXA11430000ME
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:95-69-2
- Description:4-Chloro-o-toluidine 100 µg/ml in methanol
- Formula:C₇H₈ClN
- Description:4-Chloro-o-toluidine