Heptadecafluorooctanesulphonamide
Katalog # EHERC15987110
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:499.15 g/mol
EINECS:212-046-0
CAS Number:754-91-6
Formula:C₈H₂F₁₇NO₂S
MDL Number:MFCD03094345