Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Chloro-2,5-diethoxy-4-nitrobenzene
1-Chloro-2,5-diethoxy-4-nitrobenzene
1-Chloro-2,5-diethoxy-4-nitrobenzene
1-Chloro-2,5-diethoxy-4-nitrobenzene
Supplier:
Thermo Scientific
Certificates
Expand image
1-Chloro-2,5-diethoxy-4-nitrobenzene
Supplier:
Thermo Scientific
About this item
CAS Number:
91-43-0
:
Formula:
C₁₀H₁₂ClNO₄
:
245.66 g/mol
EINECS:
202-067-3