Skip to Header
Skip to Main Content
Skip to Footer
Avansert
Avansert
SDS
Sertifikat
Produsentdata
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dimethylphenylhydrazinium chloride
3,4-Dimethylphenylhydrazinium chloride
3,4-Dimethylphenylhydrazinium chloride
3,4-Dimethylphenylhydrazinium chloride
Supplier:
Thermo Scientific
Certificates
Expand image
3,4-Dimethylphenylhydrazinium chloride
Supplier:
Thermo Scientific
About this item
CAS Number:
60481-51-8
:
MFCD00052270
Formula:
C₈H₁₃ClN₂
:
172.66 g/mol