Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-p-anisaldehyde
3-Bromo-p-anisaldehyde
3-Bromo-p-anisaldehyde
3-Bromo-p-anisaldehyde
Supplier:
Thermo Scientific
Certificates
Expand image
3-Bromo-p-anisaldehyde
Supplier:
Thermo Scientific
About this item
CAS Number:
34841-06-0
:
Formula:
C₈H₇BrO₂
MDL Number:
MFCD00016599
:
215.05 g/mol
Boiling Pt:
108…110 °C (1 mmHg)
Melting Pt:
54 °C
EINECS:
252-241-8