Order Entry
Norway
ContactUsLinkComponent
Tetrachloroethylene ≥99.5% stabilised
Tetrachloroethylene ≥99.5% stabilised
Certificates
undefined
Tetrachloroethylene ≥99.5% stabilised

Specification Test Results

  • Vapor Density:
    5.83 (vs air)
  • Contains:
    stabilizer
  • Assay:
    Min. 99.5 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Free acid (as HCl):
    Max. 0.001 %
  • Refractive Index:
    n20/D 1.505 (lit.)
  • Vapor Pressure:
    19 mmHg ( 25 °C)

Specification Test Results

Related Information