kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Tetrachloroethylene
Tetrachloroethylene ≥99%
Tetrachloroethylene ≥99%
:
Thermo Fisher Scientific
Tetrachloroethylene ≥99%
:
Thermo Fisher Scientific
:
127-18-4
:
:
C₂Cl₄
:
MFCD00000834
:
165.83 g/mol
:
14,9190
:
120…121 °C
:
–22 °C
:
1.623
:
Ambient
:
204-825-9
:
1897
:
6.1,III
:
GHS08
:
GHS09
msds_ALFAB20089
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years