Tetrachloroethylene 99+%, Spectrophotometric Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:127-18-4
:
Formula:C₂Cl₄
MDL Number:MFCD00000834
:165.83 g/mol
Merck Index:12,09332
Boiling Pt:120…122 °C (760 mmHg)
Melting Pt:–22 °C
Density:1.625 g/cm³
Storage Temperature:Ambient
EINECS:204-825-9
UN:1897
ADR:6.1,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.05 %
- GC:≥99.0 %
- UV:1 cm cell vs water
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %