
trans-4-Decenal 96% may cont. ca 7% cis-isomer
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65405-70-1
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00065447
:154.25 g/mol
Boiling Pt:80 °C (4 mmHg)
Density:0.842
Flash Pt:85 °C (185 °F)
EINECS:265-741-6
:GHS07