Order Entry
Norway
ContactUsLinkComponent
n-Tetradecane for synthesis, Sigma-Aldrich®
Health
n-Tetradecane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Health
n-Tetradecane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
     - 0.0001 g/l (20 °C)
  • Melting Point:
    6 °C
  • Molar Mass:
    198.39 g/mol
  • Boiling Point:
    254 °C
  • Vapor Pressure:
    1.55 Pa (25 °C)
  • Flash Point:
    102 °C
  • Explosion Limit:
    0.5 - 6.5 %(V)
  • Density:
    0.76 g/cm3 (20 °C)
  • Ignition Point:
    202 °C
  • Viscosity Kinematic:
    3 mm2/s (40 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together