n-Nonadecane for synthesis, Sigma-Aldrich®
: Merck
:629-92-5
:
:H₃C(CH₂)₁₇CH₃
:MFCD00009012
:268.53 g/mol
:329.7 °C (1013 hPa)
:32.5 °C
:0.7845 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:Ambient
:211-116-8
- Solubility:(20 °C) insoluble
- Melting Point:31 - 34 °C
- Molar Mass:268.52 g/mol
- Boiling Point:329 - 331 °C
- Vapor Pressure:- 0.1 hPa (20 °C)
- Flash Point:100 °C
- Refractive Index:1.4211 (70 °C, 589 nm)
- Explosion Limit:0.6 - 6.5 %(V)
- Density:0.79 g/cm3 (20 °C)
- Ignition Point:205 °C