



n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:142-82-5
:
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544
:100.20 g/mol
Merck Index:13,04679
Boiling Pt:98.4 °C (1013 hPa)
Melting Pt:–90,6 °C
Density:0.6838 g/cm³ (20 °C)
Flash Pt:–4 °C
EINECS:205-563-8
UN:1206
ADR:3,II
REACH:01-2119457603-38
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Assay:Min. 99.0 %
- IR Spectrum:Passes test
- Acidity:Max. 0.0001 meq/g
- Alkalinity:Max. 0.0001 meq/g
- Boiling point:98 - 99 °C
- Bromine value:Max. 0.5
- Colouration:Max. 10 APHA
- Density (20/4):0.681 - 0.685
- Substances coloured by H2SO4:Max. 90 APHA
- Aromatics (V/V):Max. 0.1 %
- Evaporation residue:Max. 10 ppm
- Total S (as SO4):Max. 10 ppm
- Water:Max. 100 ppm
- Al (Aluminium):Max. 0.1 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.02 ppm
- Ca (Calcium):Max. 0.1 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 0.1 ppm
- K (Potassium):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.05 ppm
- Mn (Manganese):Max. 0.02 ppm
- Na (Sodium):Max. 0.1 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.02 ppm
- Sn (Tin):Max. 0.1 ppm
- Sr (Strontium):Max. 0.02 ppm
- Zn (Zinc):Max. 0.1 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Aluminium bottle for liquids DIN 45 UN X (ZPACK8000430)
Packaging Information
spec_VWRC24551.290 spec_VWRC24551.290 spec_VWRC24551.324 spec_VWRC24551.324 spec_VWRC24551.368
Product specification
More