Order Entry
Norway
ContactUsLinkComponent
n-Heptadecane
n-Heptadecane
Catalog # PROOCHE169
CAS Number:  
n-Heptadecane
Catalog # PROOCHE169
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :240.47 g/mol
Boiling Pt:302.1 °C (1013 hPa)
Boiling Pt:302,1 °C (1013 hPa)
Melting Pt:21.2 °C
Melting Pt:21,2 °C
Density:0,7783 g/cm³ (20 °C)
Flash Pt:148 °C
Storage Temperature:Ambient
EINECS:211-108-4
 :GHS08
CAS Number:629-78-7
Formula:H₃C(CH₂)₁₅CH₃
MDL Number:MFCD00009002
Flash Pt:148 °C