Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
n-Heptadecane
n-Heptadecane ≥99.5%
n-Heptadecane ≥99.5%
44514.03
:
Thermo Fisher Scientific
:
n-Heptadecane ≥99.5%
44514.03
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
240.47 g/mol
:
302 °C
:
22 °C
:
0.7783
:
148 °C
:
Ambient
:
211-108-4
:
GHS07
:
629-78-7
:
H₃C(CH₂)₁₅CH₃
:
MFCD00009002