Order Entry
Norway
ContactUsLinkComponent
 
Sodium dodecyl sulphate (SDS) ≥98.5%, Gen-Apex for molecular biology
  33629.266
 :  
undefined
Sodium dodecyl sulphate (SDS) ≥98.5%, Gen-Apex for molecular biology
  33629.266
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Assay (calculated on anhydrous):
    Min. 98.5 %
  • Colouration (0,2 mol/l; water):
    Max. 10 APHA
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Transmittance (230 nm) (3%):
    Min. 75%
  • Transmittance (260 nm) (3%):
    Min. 80%
  • Transmittance (280 nm) (3%):
    Min. 90%
  • Transmittance (320 nm) (3%):
    Min. 95%

 

 

 :
 :288.38 g/mol
 :14,08636
 :204…207 °C
 :1.1 g/cm³ (20 °C)
 :100 °C (closed cup)
 :Ambient
 :205-788-1
 :1325
 :4.1,III
 :01-2119489461-32
 :GHS02
 :GHS07
 :GHS05
 :151-21-3
 :CH₃(CH₂)₁₁OSO₃Na
 :MFCD00036175