Order Entry
Norway
ContactUsLinkComponent
Glycerine ≥99.5%, Molecular Biology Grade
Glycerine ≥99.5%, Molecular Biology Grade
Catalog # SHETIB15760
Supplier:  IBI SCIENTIFIC
CAS Number:  
undefined
Glycerine ≥99.5%, Molecular Biology Grade
Catalog # SHETIB15760
Supplier:  IBI SCIENTIFIC
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 ml
  • Seq:
    2725

Specifications

Specification Test Results

  • Purity:
    >99.5%
  • Sulfate:
    <0.0001%
  • Arsenic:
    <0.0003%
  • Chlorine:
    <0.0003%
  • Iron :
    <0.0005%
  • Residue on Ignition:
    <0.01%
  • Density:
    1.249-1.262 g/ml
  • Specific Gravity:
    1.2635 – 1.2615
  • DNase Assay:
    None Detected
  • RNase Assay:
    None Detected
  • Protease Assay:
    None Detected

Specification Test Results

About this item

 :
 :92.09 g/mol
Merck Index:13,04497
Boiling Pt:290 °C (1013 hPa)
Melting Pt:18.6 °C
Density:1.26 g/cm³ (20 °C)
Flash Pt:177 °C
Storage Temperature:Ambient
EINECS:200-289-5
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722