Order Entry
Norway
ContactUsLinkComponent
Glycerine ≥98%, GPR RECTAPUR®
Glycerine ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Glycerine ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %
  • Density (20/4):
    1.257 - 1.265
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Fe (Iron):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 0.05 %

Specification Test Results