Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Ordreinnføring
Ordrestatus
Din konto
Handlevogn
Norway
Ordreinnføring
Ordrestatus
Din konto
Handlevogn
ContactUsLinkComponent
Home
Glycerine
Glycerine
Glycerine
MOLEM63186664
:
Molekula
:
Glycerine
MOLEM63186664
:
Molekula
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
92.09 g/mol
:
13,04497
:
290 °C (1013 hPa)
:
18.6 °C
:
1.26 g/cm³ (20 °C)
:
177 °C
:
Ambient
:
200-289-5
:
56-81-5
:
HOCH₂CH(OH)CH₂OH
:
MFCD00004722