Glycerine ≥99.5%, Ultrapure
Catalog # ICNA04800688
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Synonyms:1,2,3-Propanetriol; Glycerin; trehydroxypropane
- Form:Liquid
- Storage:Store at room temperature (15 to 30 °C)
- Heavy Metals:5 ppm (Lead)
- pH:5,0 -7,5 (1% aq soln)
- Refractive Index:n20/D 1,474 (Lit.)
Specification Test Results
About this item
:
:92.09 g/mol
Merck Index:13,04497
Boiling Pt:290 °C (1013 hPa)
Melting Pt:18.6 °C
Density:1.26 g/cm³ (20 °C)
Flash Pt:177 °C
Storage Temperature:Ambient
EINECS:200-289-5
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722