Glycerine 99.5% for molecular biology
Catalog # ACRO327255000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.5 % (Coulometric)
- GC:≥99.5 %
- UV:at 260 nm A: ≤0.06
- Color scale:≤10 APHA
- pH:6 to 7 (10 % at 25°C)
- Dnase,Rnase,Protease activity:none detected
- Heavy metals (as Pb):≤5 ppm
- Acidity (CH3COOH):≤0.003 %
- Aldehyde:≤0.001 %
Specification Test Results
About this item
:
:92.09 g/mol
Merck Index:13,04497
Boiling Pt:290 °C (1013 hPa)
Melting Pt:18 °C
Density:1.261 g/cm³
Flash Pt:160 °C
Storage Temperature:Ambient
EINECS:200-289-5
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722