


1-Propanol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71-23-8
:
Formula:H₃CCH₂CH₂OH
MDL Number:MFCD00002941
:60.1 g/mol
Merck Index:14,7842
Boiling Pt:97 °C
Melting Pt:–127 °C
Density:0.804
Flash Pt:15 °C (59 °F)
Storage Temperature:Ambient
EINECS:200-746-9
UN:1274
ADR:3,II
:GHS02
:GHS05
:GHS07