

1-Pentanol ≥98.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 20791.292
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay:Min. 98.5 %
- Boiling point:137 - 139 °C
- Density (20/4):0.813 - 0.817
- Non-volatile residue:Max. 50 ppm
- Al (Aluminium):Max. 0.5 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.1 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.02 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.1 ppm
- Sn (Tin):Max. 0.1 ppm
- Zn (Zinc):Max. 0.1 ppm
Specification Test Results
About this item
:
:88.15 g/mol
Merck Index:13,07195
Boiling Pt:137.9 °C (1013 hPa)
Melting Pt:–79 °C
Density:0.81 g/cm³ (20 °C)
Flash Pt:48 °C
Storage Temperature:Ambient
EINECS:200-752-1
UN:1105
ADR:3,III
:GHS02
:GHS07
CAS Number:71-41-0
Formula:H₃C(CH₂)₃CH₂OH
MDL Number:MFCD00002977