1-Pentanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:71-41-0
:
Formula:H₃C(CH₂)₃CH₂OH
MDL Number:MFCD00002977
:88.15 g/mol
Merck Index:13,07195
Boiling Pt:137.9 °C (1013 hPa)
Melting Pt:–79 °C
Density:0.81 g/cm³ (20 °C)
EINECS:200-752-1
UN:1105
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:48 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:22 g/l (20 °C)
- Melting Point:-79 °C
- Molar Mass:88.15 g/mol
- Boiling Point:138 °C (1013 hPa)
- Vapor Pressure:2.6 hPa (20 °C)
- Flash Point:49 °C
- Refractive Index:1.41 (20 °C, 589 nm)
- Explosion Limit:1.6 %(V)
- Density:0.815 g/cm3 (20 °C)
- pH:(22 g/l, H2O, 20 °C) neutral
- Ignition Point:300 °C DIN 51794
- Evaporation Number:2.7