Order Entry
Norway
ContactUsLinkComponent
Diklormetan, Supelco®
Diklormetan, Supelco®
Catalog # 02575-5ML
Supplier:  Merck
CAS Number:  
undefined
Diklormetan, Supelco®
Catalog # 02575-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pakketype:
    Glass bottle
  • Fp:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :84.93 g/mol
Merck Index:12,06140
Boiling Pt:39,8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1,322 g/cm³ (20 °C)
Storage Temperature:Romtemperatur
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
 :GHS08
 :GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881