Order Entry
Norway
Orders LinkContactUsLinkComponent
 

1,5-Dichloropentane ≥98%

Flammable
Irritant
1,5-Dichloropentane ≥98%
Certificates
1,5-Dichloropentane ≥98%
1,5-Dichloropentane ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 628-76-2 Molecular Formula: C5H10Cl2 Formula Weight: 141.04 Flash Point: 26°(78°F) Refractive Index: 1.4570 Melting Point: -72° Boiling Point: 182° Beilstein Reference: 1732340 UN No.: 1152 MDL No.: MFCD00001017

Synonyms:  Pentamethylendichloride
  • CAS Number:628-76-2
  • Formula:ClH₂C(CH₂)₃CH₂Cl
  • MDL Number:MFCD00001017
  • Molecular Weight:141.04 g/mol
  • Boiling Pt:182 °C
  • Melting Pt:–72 °C
  • Density:1.103
  • Flash Pt:26 °C (78 °F)
  • Storage Temperature:Ambient
  • EINECS:211-053-6
  • UN:1152
  • ADR:3,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.