
Sodium propionate for synthesis, Sigma-Aldrich®
: Merck
:137-40-6
:
:C₃H₅NaO₂
:MFCD00002759
:96.06 g/mol
:12,08816
:288.76 °C
:M-Clarity™ quality level = MQ200
:157 °C
:205-290-4
:GHS07
:Ambient
- Solubility:995 g/l (20 °C)
- Melting Point:285 - 286 °C
- Molar Mass:96.06 g/mol
- pH:8 - 9.5 (10 g/l, H2O, 20 °C)