Lignoceric acid
Katalog # EHERC17396000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:368.64 g/mol
Merck Index:13,05509
Boiling Pt:406 °C (1013 hPa)
Melting Pt:87 °C
Density:0.8207 g/cm³ (100 °C)
Storage Temperature:Ambient
EINECS:209-180-7
:GHS07
CAS Number:557-59-5
Formula:C₂₄H₄₈O₂
MDL Number:MFCD00002810