


Propionic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:79-09-4
:
Formula:C₃H₆O₂
MDL Number:MFCD00002756
:74.08 g/mol
Merck Index:13,07917
Boiling Pt:141 °C (1013 hPa)
Melting Pt:–21 °C
Density:0.99 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-176-3
UN:3463
ADR:8 (3),II
:M-Clarity™ quality level = MQ200
Flash Pt:49 °C
REACH:01-2119486971-24
:GHS02
:GHS05
:GHS07
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-20 °C
- Molar Mass:74.08 g/mol
- Boiling Point:141 °C (1013 hPa)
- Vapor Pressure:5 hPa (20 °C)
- Flash Point:50 °C
- Refractive Index:1.386 (20 °C, 589 nm)
- Explosion Limit:2.1 - 12 %(V)
- Density:0.993 g/cm3 (20 °C)
- pH:2.5 (100 g/l, H2O, 20 °C)
- Ignition Point:485 °C DIN 51794
- Synonyms:Methyl acetic acid
- Hill Formula:C3H6O2