Propionic acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-09-4
:
Formula:C₃H₆O₂
MDL Number:MFCD00002756
:74.08 g/mol
Merck Index:14,7825
Boiling Pt:140…141 °C
Melting Pt:–21 °C
Density:0.992
Flash Pt:51 °C (123 °F)
Storage Temperature:Ambient
EINECS:201-176-3
UN:3463
ADR:8,II
REACH:01-2119486971-24
:GHS05
:GHS02
Related Information
Certificates
Search by Lot NumberALFAL04210.22_TDS_EN
Product specification
Organic Building Blocks and Intermediates