Palmitic acid
Catalog # EHERC14191800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
Merck Index:13,07064
Boiling Pt:352 °C (1013 hPa)
Melting Pt:61…63 °C
Density:0.853 g/cm³ (20 °C)
:256.43 g/mol
Storage Temperature:Ambient
EINECS:200-312-9
CAS Number:57-10-3
Formula:C₁₆H₃₂O₂
MDL Number:MFCD00002747