Margaric acid
Catalog # EHERC14122600
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:270.46 g/mol
Merck Index:13,05775
Boiling Pt:364 °C (1013 hPa)
Melting Pt:63 °C
Density:1.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-027-1
:GHS07
CAS Number:506-12-7
Formula:C₁₇H₃₄O₂
MDL Number:MFCD00002751