
Heneicosanoic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2363-71-5
:
Formula:C₂₁H₄₂O₂
MDL Number:MFCD00002805
:326.56 g/mol
Boiling Pt:170 °C (0.1 mbar)
Melting Pt:74…76 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:219-113-3
:GHS07
Specification Test Results
- Appearance:White Glistening crystalline powder
- Infrared spectrum:Conforms
- GC:≥98.5 %